2-Thiobarbituric acid 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine
- CAS Number:504-17-6
- Formula:C₄H₄N₂O₂S
- MDL Number:MFCD00006674
- Molecular Weight:144.15 g/mol
- Boiling Pt:419 °C (1013 hPa)
- Melting Pt:240…255 °C
- Density:1,661 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-985-8
3 Options Available Below
Product Details & Documents
Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine
- CAS Number:504-17-6
- Formula:C₄H₄N₂O₂S
- MDL Number:MFCD00006674
- Molecular Weight:144.15 g/mol
- Boiling Pt:419 °C (1013 hPa)
- Melting Pt:240…255 °C
- Density:1,661 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-985-8
Specifications for Product Family
Specification Test Results
- Appearance:White to yellow to light brown Powder or crystalline solid
- Infrared spectrum:Conforms
- Solubility:(5% in 1M NaOH) Clear to slightly hazy
- HPLC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %



