Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chrysene
Chrysene
Chrysene
123002H
:
VWR Chemicals
:
Chrysene
123002H
:
VWR Chemicals
:
Pack type:
Glass ampoule
Pk:
100 mg
Seq:
2440
:
:
228.29 g/mol
:
13,02275
:
448 °C (1013 hPa)
:
250…252 °C
:
1.274 g/cm³ (20 °C)
:
205-923-4
:
2811
:
6.1,III
:
GHS08
:
GHS09
:
218-01-9
:
C₁₈H₁₂
:
MFCD00003698