Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzo[e]pyrene

Benzo[e]pyrene

Glass bottle for solids, 100 mg
Catalog # PROOBCR-050
Health
Environment
Benzo[e]pyrene
Catalog # PROOBCR-050
CAS Number:  192-97-2
Benzo[e]pyrene

Glass bottle for solids, 100 mg

CAS Number:  192-97-2
Price
7.310,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  BeP
  • CAS Number:192-97-2
  • Formula:C₂₀H₁₂
  • Molecular Weight:252.32 g/mol
  • Merck Index:12,01135
  • Boiling Pt:492 °C (1013 hPa)
  • Melting Pt:178…179 °C
  • EINECS:205-892-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Cat. No.:
    PROOBCR-050

Specifications