4-Chlorobiphenyl (PCB No. 3)
Catalog # EHERC20000300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
Boiling Pt:291 °C (1013 hPa)
Melting Pt:77…78 °C
:188.66 g/mol
EINECS:218-127-7
CAS Number:2051-62-9
Formula:C₁₂H₉Cl
MDL Number:MFCD00013640