Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymol blue

Thymol blue

100
Catalog # HACH2067299
Supplier:   Hach
Thymol blue
Catalog # HACH2067299
Supplier:  Hach
CAS Number:  76-61-9
Thymol blue

100

CAS Number:  76-61-9
Price
445,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Melting Pt:221 °C
  • Storage Temperature:Ambient
  • EINECS:200-973-3
Specifications

Specifications

  • Pk:
    100

Specifications