Bromophenol blue
Glass bottle for solids, 5 g
Catalog # ROTHT116.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Number:115-39-9
- Formula:C₁₉H₁₀Br₄O₅S
- MDL Number:MFCD00005875
- Molecular Weight:669.97 g/mol
- Merck Index:13,01429
- Melting Pt:273 °C
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-086-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



