Order Entry
Denmark
Orders LinkContactUsLinkComponent
Toluene

Toluene

Glass bottle, 2,5 L
Catalog # ROTHT166.1
Flammable
Health
Irritant
Marketsource
Supplier:   Roth Carl
Toluene
Catalog # ROTHT166.1
Supplier:  Roth Carl
Marketsource
CAS Number:  108-88-3
Toluene
Toluene

Glass bottle, 2,5 L

CAS Number:  108-88-3
Price
589,20 kr
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Methylbenzene, Phenylmethane, FM approval
  • CAS Number:108-88-3
  • Formula:C₆H₅CH₃
  • MDL Number:MFCD00008512
  • Molecular Weight:92.14 g/mol
  • Merck Index:13,09607
  • Boiling Pt:110.6 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:0.867 g/cm³ (20 °C)
  • Flash Pt:4 °C
  • Storage Temperature:Ambient
  • EINECS:203-625-9
  • UN:1294
  • ADR:3,II
  • REACH:05-2114615130-69
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.