Order Entry
Denmark
ContactUsLinkComponent
3,5-Dihydroxytoluene for synthesis, Sigma-Aldrich®
3,5-Dihydroxytoluene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
3,5-Dihydroxytoluene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    80 g/l (20 °C)
  • Melting Point:
    109 °C
  • Molar Mass:
    124.14 g/mol
  • Boiling Point:
    147 °C (7 hPa)
  • Flash Point:
    159 °C

Specification Test Results

Related Information

Frequently Bought Together