

3,5-Dihydroxytoluene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-15-4
:
Formula:C₇H₈O₂
MDL Number:MFCD00002291
:124.14 g/mol
Merck Index:14,6864
Boiling Pt:287…290 °C
Melting Pt:106…108 °C
Density:1.2895 g/cm³ (20 °C)
Flash Pt:159 °C
Storage Temperature:Ambient
EINECS:207-984-2
:GHS06
:GHS07
Related Information
Frequently Bought Together