
Benzyl alcohol, EMSURE® for analysis, Supelco®
: Merck
:100-51-6
:
:C₆H₅CH₂OH
:MFCD00004599
:108.14 g/mol
:13,01126
:205 °C (1013 hPa)
:–15.3 °C
:1.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:101 °C
:202-859-9
:GHS07
:Ambient
- Solubility:40 g/l (20 °C)
- Melting Point:-15.3 °C
- Molar Mass:108.14 g/mol
- Boiling Point:205 °C (1013 hPa)
- Vapor Pressure:0.03 hPa (20 °C)
- Flash Point:101 °C DIN 51758
- Refractive Index:1.54
- Explosion Limit:1.3 - 13 %(V)
- Density:1.05 g/cm3 (20 °C)
- Ignition Point:435 °C DIN 51794
Frequently Bought Together