Ordreindtastning
Denmark
ContactUsLinkComponent
 
Benzylalkohol 98.0-100.5% (by GC), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
 
undefined
Benzylalkohol 98.0-100.5% (by GC), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
 

 

  • Assay (GC):
    98.0 - 100.5%
  • Identity (IR):
    Conforms
  • Acidity:
    Conforms
  • Refractive Index (n 20/D):
    1.538 - 1.541
  • Density (d 20 °C/20 °C):
    1.043 - 1.049
  • Peroxide Value:
    ≤5.0
  • Related Substances (GC) (Benzaldehyde (Impurity A)):
    ≤0.15%
  • Related Substances (GC) (Cyclohexylmethanol (Impurity B)):
    ≤0.10%
  • Related Substances (GC) (Sum of Peaks with a Relative Retention Less than that of Benzyl Alcohol):
    ≤0.04%
  • Related Substances (GC) (Sum of Peaks with a Relative Retention Greater than that of Benzyl Alcohol):
    ≤0.3%
  • Related Substances (GC) (Disregard Limit):
    ≤0.0001%
  • Benzene (GC):
    ≤0.0002%
  • Chlorobenzene (GC):
    ≤0.01%
  • Toluene (GC):
    ≤0.01%
  • Other Residual Solvents (ICH Q3C):
    Excluded by Manufacturing Process
  • Evaporation Residue:
    ≤0.050%
  • Water:
    ≤0.10%

 

Frequently Bought Together