Eugenol
Glass bottle for solids, 1 g
Catalog # ROTH6941.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:13,03931
- Boiling Pt:255 °C (1013 hPa)
- Melting Pt:∼–10 °C
- Density:1.0652 g/cm³ (20 °C)
- Flash Pt:110 °C
- Storage Temperature:Ambient
- EINECS:202-589-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



