Order Entry
Denmark
Orders LinkContactUsLinkComponent
Eugenol ≥99%
Eugenol ≥99%
Eugenol ≥99%
Eugenol ≥99%
Purity: 99% CAS No.: 97-53-0 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 127°(260°F) Refractive Index: 1.5410 Odour: Pungent Melting Point: -10° Boiling Point: 252-253° Beilstein Reference: 1366759 Merck Reference: 14,3898 MDL No.: MFCD00008654
 :  4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
  •  :97-53-0
  •  :C₁₀H₁₂O₂
  •  :MFCD00008654
  •  :164.2 g/mol
  •  :14,3898
  •  :252…253 °C
  •  :–10 °C
  •  :1.068
  •  :127 °C (260 °F)
  •  :Ambient
  •  :202-589-1
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 471,00 kr
    471,00 kr
    1 * 100 g
       471,00 kr
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 1.650,00 kr
    1.650,00 kr
    1 * 500 g
       1.650,00 kr