Eugenol 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:13,03931
- Boiling Pt:254 °C (760 mmHg)
- Melting Pt:12…10 °C
- Density:1.06 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-589-1
4 Options Available Below
Product Details & Documents
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:13,03931
- Boiling Pt:254 °C (760 mmHg)
- Melting Pt:12…10 °C
- Density:1.06 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-589-1
Documents
ACRO11911
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5400 to 1.5420 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(20°C) 1.064 to 1.070
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



