Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzene

Benzene

Flammable
Health
Marketsource
Supplier:   Roth Carl
Benzene
Supplier:  Roth Carl
Marketsource
Certificates
Benzene

4 Options Available Below

Product Details & Documents
Synonyms:  Cyclohexatriene
  • CAS Number:71-43-2
  • Formula:C₆H₆
  • MDL Number:MFCD00003009
  • Molecular Weight:78.11 g/mol
  • Merck Index:13,01066
  • Boiling Pt:80.1 °C (1013 hPa)
  • Melting Pt:5.5 °C
  • Density:0.878 g/cm³ (20 °C)
  • Flash Pt:–11 °C
  • Storage Temperature:Ambient
  • EINECS:200-753-7
  • UN:1114
  • ADR:3,II
  • REACH:01-2119447106-44
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 238,22 kr
    238,22 kr
    1 * 100 mL
       238,22 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 391,30 kr
    391,30 kr
    1 * 1 L
       391,30 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
    Marketsource
    Marketsource
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.