Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Pyrogallol ≥98%

Health
Irritant
Pyrogallol ≥98%
Certificates
Pyrogallol ≥98%
Pyrogallol ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 87-66-1 Molecular Formula: C6H6O3 Formula Weight: 126.11 Odour: Odourless Melting Point: 131-135° Boiling Point: 309° Beilstein Reference: 907431 Merck Reference: 14,8000 UN No.: 2811 MDL No.: MFCD00002192

Synonyms:  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  • CAS Number:87-66-1
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00002192
  • Molecular Weight:126.11 g/mol
  • Merck Index:14,8000
  • Boiling Pt:309 °C
  • Melting Pt:131…135 °C
  • Density:1.45
  • Storage Temperature:Ambient
  • EINECS:201-762-9
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.