Pyrogallol ACS
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131.0 to 135.0 °C
- Residue after ignition:≤0.005 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤5 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



