Pyrogallol 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 135 °C
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Arsenic (As):≤0.5 ppm
- Calcium (Ca):≤20 ppm
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤5 ppm
- Potassium (K):≤20 ppm
- Heavy metals (as Pb):≤5 ppm
- Nitrogen compounds (as N):≤5 ppm



