Order Entry
Denmark
Orders LinkContactUsLinkComponent
Resorcinol

Resorcinol

Irritant
Environment
Marketsource
Supplier:   Roth Carl
Resorcinol
Supplier:  Roth Carl
Marketsource
Certificates
Resorcinol

2 Options Available Below

Product Details & Documents
Synonyms:  1,3-Dihydroxybenzene, 1,3-Benzenediol
  • CAS Number:108-46-3
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002269
  • Molecular Weight:110.11 g/mol
  • Merck Index:12,08323
  • Boiling Pt:281 °C (1013 hPa)
  • Melting Pt:109…111 °C
  • Density:1.272 g/cm³ (20 °C)
  • Flash Pt:127 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-585-2
  • UN:2876
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource