

Resorcinol 99.0-100.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 27378.234
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:99.0 - 100.5 %
- Acidity or alkalinity:Max. 0.005 meq/g
- Melting point:109 - 112 °C
- Ignition residue (SO4):Max. 0.02 %
- Insolubility in water:Max. 50 ppm
- Insolubility in methanol:Max. 50 ppm
- Phenol:0.02%
- Pyrocatechol:0.02%
Specification Test Results
About this item
:
Merck Index:12,08323
Boiling Pt:281 °C (1013 hPa)
Melting Pt:109…111 °C
Density:1.272 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-585-2
UN:2876
ADR:6.1,III
:110.11 g/mol
:GHS07
:GHS09
CAS Number:108-46-3
Formula:C₆H₆O₂
MDL Number:MFCD00002269