Order Entry
Denmark
Orders LinkContactUsLinkComponent
Resorcinol ≥99%

Resorcinol ≥99%

Irritant
Environment
Resorcinol ≥99%
Certificates
Resorcinol ≥99%
Resorcinol ≥99%

4 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 108-46-3 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 171°(339°F) Odour: Phenol-like pH: 4-6 Melting Point: 109-112° Boiling Point: 280-281° Beilstein Reference: 906905 Merck Reference: 14,8155 UN No.: 2876 MDL No.: MFCD00002269

Synonyms:  1,3-Dihydroxybenzene, 1,3-Benzenediol
  • CAS Number:108-46-3
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002269
  • Molecular Weight:110.11 g/mol
  • Merck Index:14,8155
  • Boiling Pt:280…281 °C
  • Melting Pt:109…112 °C
  • Density:1.272
  • Flash Pt:171 °C (339 °F)
  • Storage Temperature:Ambient
  • EINECS:203-585-2
  • UN:2876
  • ADR:6.1,III
Product Family Options

Product Information