Order Entry
Denmark
Orders LinkContactUsLinkComponent
Resorcinol 98%

Resorcinol 98%

Irritant
Environment
Resorcinol 98%
Certificates
Resorcinol 98%

4 Options Available Below

Product Details & Documents
Synonyms:  1,3-Dihydroxybenzene, 1,3-Benzenediol
  • CAS Number:108-46-3
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002269
  • Molecular Weight:110.11 g/mol
  • Merck Index:12,08323
  • Boiling Pt:281 °C (760 mmHg)
  • Melting Pt:109…111 °C
  • Density:1,272 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Ambient
  • EINECS:203-585-2
  • UN:2876
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow to beige to pink Crystals or granules or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    108 to 112 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.