Order Entry
Denmark
ContactUsLinkComponent
General purpose gloves, A130 Criss Cross
General purpose gloves, A130 Criss Cross
Supplier:  Portwest
Certificates
undefined
General purpose gloves, A130 Criss Cross
Supplier:  Portwest
Specifications
  • Catalog No:
  • PRTWA130ORRXL
  • PRTWA130ORRL
  • PRTWA130ORRM
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Finish:
  • PVC criss cross pattern
  • PVC criss cross pattern
  • PVC criss cross pattern
  • Gauge:
  • 7
  • 7
  • 7
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (1141X)
  • EN 388:2016 (1141X)
  • EN 388:2016 (1141X)