Thiomersal
Catalog # A1278.0100
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:404.82 g/mol
Merck Index:12,09451
Melting Pt:232…233 °C
Flash Pt:250 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-210-4
UN:2025
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:54-64-8
Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
MDL Number:MFCD00013062