Sodium salicylate
Catalog # MOLE57802768-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:160.10 g/mol
Merck Index:12,08819
Melting Pt:200 °C
Density:0.32 g/cm³ (20 °C)
EINECS:200-198-0
:GHS07
Storage Temperature:Ambient
CAS Number:54-21-7
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440