
Sodium salicylate, Millipore®
Catalog # 567630-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:160.10 g/mol
Merck Index:12,08819
Melting Pt:200 °C
Density:0.32 g/cm³ (20 °C)
EINECS:200-198-0
:GHS07
Storage Temperature:Ambient
CAS Number:54-21-7
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440