
Sodium salicylate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54-21-7
:
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440
:160.10 g/mol
Merck Index:14,8332
Melting Pt:>300 °C
Density:0.32 g/cm³ (20 °C)
EINECS:200-198-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together