Order Entry
Denmark
ContactUsLinkComponent
N-phenylanthranilic acid 98%
Irritant
N-phenylanthranilic acid 98%
Certificates
undefined
Irritant
N-phenylanthranilic acid 98%

Specification Test Results

  • Appearance:
    Yellow-green to grey-green Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    182 to 187 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together