
Propyl-4-hydroxybenzoate
Catalog # EHERC14229200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:180.2 g/mol
Merck Index:13,07958
Boiling Pt:292 °C (1013 hPa)
Melting Pt:95 °C
Density:1.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-307-7
:GHS07
CAS Number:94-13-3
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00002354