Propyl-4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
CAS Number:94-13-3
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00002354
Boiling Pt:292
Melting Pt:95
Density:1.28 g/cm³ (20 °C)
Molecular Weight:180.2 g/mol
Merck Index:13,07958
EINECS:202-307-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:98.0 - 102.0 %
- NF - Assay (C₁₀H₁₂O₃):98.0 - 102.0 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Color of Solution:Passes Test
- NF - Acidity:Passes Test
- NF - Related Substances - p-Hydroxybenzoic Acid:≤0.5 %
- NF - Related Substances - Unspecified Impurities (each):≤0.5 %
- NF - Related Substances - Total Impurities:≤1.0 %
- NF - Residue on Ignition:≤0.1 %
- EP/BP - Assay (C₁₀H₁₂O₃):98.0 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Related Substances - Impurity A:≤0.5 %
- EP/BP - Related Substances - Unspecified Impurities (each):≤0.5 %
- EP/BP - Related Substances - Total Impurities:≤1.0 %
- EP/BP - Sulfated Ash:≤0.1 %
- JP - Assay:98.0 - 102.0
- JP - Identification:Passes Test
- JP - Melting Point:96 - 99 °C
- JP - Acidity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals:≤20 ppm
- JP-RelatedSubstances- p-Hydroxybenzoic Acid:≤0.5 %
- JP-Related Substances-Unspecified Impurities (each):≤0.5 %
- JP-Related Substances-Total Impurities:≤1.0 %
- JP - Residue on Ignition:≤0.10 %
Specification Test Results
Related Information
Frequently Bought Together