Propyl-4-hydroxybenzoate 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
- CAS Number:94-13-3
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD00002354
- Molecular Weight:180.2 g/mol
- Merck Index:13,07958
- Boiling Pt:292 °C (1013 hPa)
- Melting Pt:95…99 °C
- Density:1,28 g/cm³ (20 °C)
- Flash Pt:180 °C
- Storage Temperature:Ambient
- EINECS:202-307-7
4 Options Available Below
Product Details & Documents
Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
- CAS Number:94-13-3
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD00002354
- Molecular Weight:180.2 g/mol
- Merck Index:13,07958
- Boiling Pt:292 °C (1013 hPa)
- Melting Pt:95…99 °C
- Density:1,28 g/cm³ (20 °C)
- Flash Pt:180 °C
- Storage Temperature:Ambient
- EINECS:202-307-7
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:95 to 99 °C
- Sulfated ash:≤0.1 %
- GC:≥99.0 %
- Appearance of solution:(10% in ethanol 96%) clear colorless to
Product Family Options
Product Information
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



