Order Entry
Denmark
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate

Methyl-4-hydroxybenzoate

1 g
Catalog # 47889.
Irritant
Supplier:   Merck
Methyl-4-hydroxybenzoate
Catalog # 47889.
Supplier:  Merck
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate
Methyl-4-hydroxybenzoate

1 g

CAS Number:  99-76-3
Price
190,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,06129
  • Boiling Pt:270…280 °C (1013 hPa)
  • Melting Pt:125…128 °C
  • Density:1.1208 g/cm³ (137 °C)
  • Storage Temperature:Ambient
  • EINECS:202-785-7
Specifications

Specifications

  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.