Methyl-4-hydroxybenzoate
Glass bottle for solids, 1 g
Catalog # ROTH4748.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Merck Index:13,06129
- Boiling Pt:270…280 °C (1013 hPa)
- Melting Pt:125…128 °C
- Density:1.1208 g/cm³ (137 °C)
- Storage Temperature:Ambient
- EINECS:202-785-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



