Methyl-4-hydroxybenzoate
EHERC14229000
:
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
:
:152.15 g/mol
:13,06129
:270…280 °C (1013 hPa)
:125…128 °C
:1.1208 g/cm³ (137 °C)
:Ambient
:202-785-7
:GHS07
:99-76-3
:HOC₆H₄CO₂CH₃
:MFCD00002352