Order Entry
Denmark
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate ≥99%

Methyl-4-hydroxybenzoate ≥99%

Methyl-4-hydroxybenzoate ≥99%
Certificates
Methyl-4-hydroxybenzoate ≥99%
Methyl-4-hydroxybenzoate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 99-76-3 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless pH: 5.8 Melting Point: 125-128° Boiling Point: ca 280° dec. Beilstein Reference: 509801 Merck Reference: 14,6107 MDL No.: MFCD00002352

Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,6107
  • Boiling Pt:∼280 °C (dec.)
  • Melting Pt:125…128 °C
  • Density:1.1208 g/cm³ (137 °C)
  • Storage Temperature:Ambient
  • EINECS:202-785-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.