Order Entry
Denmark
ContactUsLinkComponent
 
Methyl anthranilate
Catalog # EHERC10171470
Supplier:  EHRENSTORFER
CAS Number:  
Methyl anthranilate
Catalog # EHERC10171470
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :151.16 g/mol
Merck Index:12,06099
Boiling Pt:237 °C (1013 hPa)
Melting Pt:24 °C
Density:1.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-132-4
CAS Number:134-20-3
Formula:C₈H₉NO₂
MDL Number:MFCD00007710