Order Entry
Denmark
ContactUsLinkComponent
5-Chloro-2-fluorobenzoic acid
5-Chloro-2-fluorobenzoic acid
Catalog # MOLE19115143-5G
Supplier:  Molekula
CAS Number:  
5-Chloro-2-fluorobenzoic acid
Catalog # MOLE19115143-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

 :
 :174.56 g/mol
 :GHS07
CAS Number:394-30-9
Formula:C₇H₄ClFO₂
MDL Number:MFCD00665762