Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Vanillinsyre

Glass bottle for solids, 10 g
Catalog # ROTH3685.1
Marketsource
Supplier:   Roth Carl
Vanillinsyre
Catalog # ROTH3685.1
Supplier:  Roth Carl
Marketsource
CAS Number:  121-34-6
Vanillinsyre

Glass bottle for solids, 10 g

CAS Number:  121-34-6
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzoic acid
  • CAS Number:121-34-6
  • Formula:C₈H₈O₄
  • MDL Number:MFCD00002551
  • Molecular Weight:168.15 g/mol
  • Merck Index:12,10068
  • Boiling Pt:353 °C (1013 hPa)
  • Storage Temperature:Stue temperatur
  • EINECS:204-466-8
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifications