4-Methylmorpholine 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: NMM, N-Methylmorpholine
- CAS Number:109-02-4
- Formula:C₅H₁₁NO
- MDL Number:MFCD00006175
- Molecular Weight:101.15 g/mol
- Boiling Pt:115…116 °C (750 mmHg)
- Melting Pt:–65 °C
- Density:0.92 g/cm³
- Flash Pt:14 °C
- Storage Temperature:Ambient
- EINECS:203-640-0
- UN:2535
- ADR:3,II
4 Options Available Below
Product Details & Documents
Synonyms: NMM, N-Methylmorpholine
- CAS Number:109-02-4
- Formula:C₅H₁₁NO
- MDL Number:MFCD00006175
- Molecular Weight:101.15 g/mol
- Boiling Pt:115…116 °C (750 mmHg)
- Melting Pt:–65 °C
- Density:0.92 g/cm³
- Flash Pt:14 °C
- Storage Temperature:Ambient
- EINECS:203-640-0
- UN:2535
- ADR:3,II
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4339 to 1.4359 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥98.5 %
- Color scale:≤50 APHA
Product Family Options
Product Information
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



