Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cycloheximide

Cycloheximide

Plastic bottle for solids, 500 mg
Catalog # USBIC8500
Toxic
Health
Environment
Supplier:   US Biological
Cycloheximide
Catalog # USBIC8500
Supplier:  US Biological
CAS Number:  66-81-9
Cycloheximide
Cycloheximide

Plastic bottle for solids, 500 mg

CAS Number:  66-81-9
Price
526,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.