

2-Butyne-1-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:764-01-2
:
Formula:C₄H₆O
MDL Number:MFCD00002914
:70.09 g/mol
Boiling Pt:142…143 °C
Melting Pt:–2 °C
Density:0.938
Flash Pt:51 °C (123 °F)
Storage Temperature:Ambient
EINECS:212-113-4
UN:1987
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together