2-Butyne-1-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:764-01-2
:
Formula:C₄H₆O
MDL Number:MFCD00002914
:70.09 g/mol
Boiling Pt:142 °C (1013 hPa)
Melting Pt:–2.2 °C
Density:0.93 g/cm³
Flash Pt:51 °C
Storage Temperature:Ambient
EINECS:212-113-4
UN:1987
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4520 to 1.4540 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together