Order Entry
Denmark
Orders LinkContactUsLinkComponent
Caffeic acid, Supelco®

Caffeic acid, Supelco®

Glass bottle for solids, 1 g
Catalog # 60018-1G
Supplier:   Merck
Caffeic acid, Supelco®
Catalog # 60018-1G
Supplier:  Merck
CAS Number:  331-39-5
Caffeic acid, Supelco®
Caffeic acid, Supelco®

Glass bottle for solids, 1 g

CAS Number:  331-39-5
Price
731,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Merck Index:12,01673
  • Boiling Pt:366 °C (1013 hPa)
  • Melting Pt:234…237 °C
  •  :M-Clarity™ quality level = MQ100
  • EINECS:206-361-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications