Order Entry
Denmark
ContactUsLinkComponent
Dimethyl fumarate 99%
Dimethyl fumarate 99%
Certificates
undefined
Dimethyl fumarate 99%
Certificates

Specification Test Results

  • Appearance:
    White to light beige to light brown Crystalline powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    100 to 107 °C
  • GC:
    ≥98.5 %
  • Free acid:
    ≤1.5 % (as Fumaric acid)

Specification Test Results

Related Information

Frequently Bought Together