Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bisacrylamide ≥97%

Bisacrylamide ≥97%

Irritant
Leverandør:   Thermo Fisher Scientific
Bisacrylamide ≥97%
Certificates
Bisacrylamide ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625

Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:125 °C (3.33 hPa)
  • Melting Pt:>300 °C
  • Density:1.235
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Product Family Options

Product Information

  • Pk
    Tilgængelighed
    Price
  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 547,00 kr
    547,00 kr
    1 * 100 g
       547,00 kr

  • 500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1.650,00 kr
    1.650,00 kr
    1 * 500 g
       1.650,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.