Bisacrylamide ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99+% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Physical Form: Powder Density: 1.235 Appearance: White Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625
Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Number:110-26-9
- Formula:(H₂C=CHCONH)₂CH₂
- MDL Number:MFCD00008625
- Molecular Weight:154.17 g/mol
- Boiling Pt:125 °C (3.33 hPa)
- Melting Pt:>300 °C
- Density:1.235
- Flash Pt:138 °C
- Storage Temperature:Ambient
- EINECS:203-750-9
2 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Physical Form: Powder Density: 1.235 Appearance: White Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625
Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Number:110-26-9
- Formula:(H₂C=CHCONH)₂CH₂
- MDL Number:MFCD00008625
- Molecular Weight:154.17 g/mol
- Boiling Pt:125 °C (3.33 hPa)
- Melting Pt:>300 °C
- Density:1.235
- Flash Pt:138 °C
- Storage Temperature:Ambient
- EINECS:203-750-9



