Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acrylamide, 4x crystallised

Acrylamide, 4x crystallised

Toxic
Health
Marketsource
Supplier:   Serva
Acrylamide, 4x crystallised
Supplier:  Serva
Marketsource
Certificates
Acrylamide, 4x crystallised
Acrylamide, 4x crystallised

2 Options Available Below

Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 395,78 kr
    395,78 kr
    1 * 100 g
       395,78 kr

    Specifications:
    Marketsource
    Marketsource
  • Plastic bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.