Acrylamide, 2x crystallised
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:84.5 °C
- Density:1.122 g/cm³ (20 °C)
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:84.5 °C
- Density:1.122 g/cm³ (20 °C)
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



