Order Entry
Denmark
ContactUsLinkComponent
cis-4-Heptenal, Supelco®
cis-4-Heptenal, Supelco®
Catalog # 00931-5ML
Supplier:  Merck
CAS Number:  
undefined
cis-4-Heptenal, Supelco®
Catalog # 00931-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :112.17 g/mol
Boiling Pt:46 °C (1013 hPa)
Density:0.85 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,III
 :M-Clarity™ quality level = MQ100
EINECS:229-779-7
UN:1989
CAS Number:6728-31-0
Formula:C₇H₁₂O
MDL Number:MFCD00014682