Isoamyl alcohol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 8010.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (by GC):min. 98 %
- Acids and Esters (as Amyl Acetate):max. 0.06 %
- Aldehydes:passes test
- Boiling Range:128 - 132 °C
- Color (APHA):max. 5
- Residue after Evaporation:max. 0.003 %
- Substances Darkened by H2SO4:passes test
- Titrable Acid (meq/g):max. 0.002
- Water (H2O):max. 0.2 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
About this item
Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
:
:88.15 g/mol
Melting Pt:–117 °C
EINECS:204-633-5
UN:1105
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:123-51-3
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934
Boiling Pt:130
Density:0.812 g/cm³ (20 °C)
Flash Pt:42
ADR:3 III
Merck Index:13,05214